Find methods for your needs
Improved extraction of substituted phenethylamines from urine using Thermo Scientific HyperSep Servo+ AMP SPE and a Thermo Scientific TraceGOLD TG-5MS GC column
Description
The Thermo Scientific GC system is applied for the analysis of commonly abused substituted phenethylamines from urine. The samples are prepared using Thermo Scientific HyperSep Servo+ AMP SPE cartridges, and the separation is performed on a Thermo Scientific TraceGOLD TG-5MS column, with MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Clinical Research |
| Keywords: | Amphetamine, GC-MS, HyperSep Servo+ AMP, MDA, MDMA, Methamphetamine, Solid phase extraction, SPE, TraceGOLD TG-5MS, Urine |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.