Find methods for your needs
Improved identification of bath salts using Thermo Scientific HyperSep Servo+ Total B SPE and a Thermo Scientific TraceGOLD TG-5MS GC column
Description
The Thermo Scientific GC system is applied for the analysis of bath salts. The samples are prepared using Thermo Scientific HyperSep Servo+ Total B SPE cartridges, and the separation is performed on a Thermo Scientific TraceGOLD TG-5MS column, with MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Clinical Research |
| Keywords: | Bath salts, GC, HyperSep Servo+ Total B, MDPV, Mephedrone, MS detection, Solid phase extraction, SPE, TraceGOLD TG-5MS |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.