Find methods for your needs
Improved reversed-phase analysis of a monoclonal antibody using a Thermo Scientific Accucore 150-C4 HPLC column
Description
The Thermo Scientific UltiMate 3000 Titanium HPLC system is applied for the reversed-phase analysis of a monoclonal antibody. The separation is performed on a Thermo Scientific Accucore 150-C4 HPLC column, with UV detection at 214 nm.| Market: | BioPharma |
| Keywords: | Accucore 150-C4, Alemtuzumab, Campath, HPLC, MAb, Monoclonal antibody |
| Matrix: | Campath monoclonal antibody |
For Research Use Only. Not for use in diagnostic procedures.