Find methods for your needs
High resolution analysis of 2-aminobenzamide labeled dextran ladder on a Thermo Scientific Accucore 150-Amide-HILIC nanoLC column
# | Compound Name | Compound Class | RT (min) |
---|---|---|---|
1 | Peak 1 | Glycan | 22.03 |
2 | Peak 2 | Glycan | 27.74 |
3 | Peak 3 | Glycan | 32.37 |
4 | Peak 4 | Glycan | 36.05 |
5 | Peak 5 | Glycan | 39.09 |
6 | Peak 6 | Glycan | 41.65 |
7 | Peak 7 | Glycan | 43.88 |
8 | Peak 8 | Glycan | 45.82 |
9 | Peak 9 | Glycan | 47.55 |
10 | Peak 10 | Glycan | 49.12 |
11 | Peak 11 | Glycan | 50.42 |
12 | Peak 12 | Glycan | 51.93 |
13 | Peak 13 | Glycan | 52.89 |
14 | Peak 14 | Glycan | 54.01 |
15 | Peak 15 | Glycan | 55.21 |
16 | Peak 16 | Glycan | 56.33 |
17 | Peak 17 | Glycan | 57.02 |
18 | Peak 18 | Glycan | 57.68 |
Description
The Thermo Scientific nanoLC system is applied for the analysis of 2-aminobenzamide labeled dextran ladder. The separation is performed on a Thermo Scientific Accucore 150-Amide-HILIC nanoLC column, with UV detection at 240 and 330 nm.Market: | Proteomics |
Keywords: | 2-aminobenzamide labeled dextran ladder, Accucore 150-Amide-HILIC, Glycans, HPLC, nanoLC, Proteomics, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.