Find methods for your needs
AN282: Rapid and Sensitive Determination of Biofuel Sugars by Ion Chromatography
Description
This work shows a rapid (<8 min) and robust method with minimal sample pretreatment using high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD) to determine the sugars of interest in the highly concentrated samples encountered in biofuel processing.| Market: | Chemical |
| Keywords: | arabinose, biofuel, carbohydrate, corn stover, Electrochemical detection, fucose, Glucose, xylose, Mannose, Sucrose, PAD, Cellobiose, fructose, CarboPac SA10, monosaccharide, galactose, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 282 |
| Matrix: | Corn stover (acid hydrolyzed) |
| Author: | Lipika Basumallick and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.