Find methods for your needs
AN1022:Determination of Voriconazole Related Compound F in Voriconazole Using IC
Description
The USP method, which specifies the Thermo Scientific Dionex OmniPac PAX-100 Analytical Column (4 × 250 mm) with manually prepared eluent and chemically suppressed conductivity detection, was evaluated for the determination of voriconazole related compound F with a threshold amount of no more than 5 μg/mL (0.1%) in a 5 mg/mL solution of voriconazole, which is the final concentration amount for intravenous (IV) administration. The method was found to successfully determine voriconazole related compound F as required by the proposed USP method.| Market: | Pharma |
| Keywords: | (1R)-(−)-10-camphorsulfonic acid, antifungal, Dionex OmniPac PAX-100 Column, intravenous, Pharmaceuticals, Triazole, voriconazole, voriconazole related compound F |
| Matrix: | Intravenous drug |
For Research Use Only. Not for use in diagnostic procedures.