Find methods for your needs
AN276: Direct Determination of Fluoroacetate in Water by IC-MS Be the first to rate this application
fluoroacetate water
Description
This study undertook an investigation to quantify fluoroacetate, a metabolic poison, in fortified drinking water without prior derivatization or sample pretreatment. An ion chromatography (IC) method was developed with an MS detector.
Market: Environmental
Keywords: drinking water, ESI, fluoroacetic acid, MSQ Plus, RFIC, Rodenticide, SIM, single quad MS
Matrix: Water
Affiliation: Thermo Fisher Scientific
Uploaded on 2/4/2015.
For Research Use Only. Not for use in diagnostic procedures.