Find methods for your needs
AN276: Direct Determination of Fluoroacetate in Water by IC-MS
Description
This study undertook an investigation to quantify fluoroacetate, a metabolic poison, in fortified drinking water without prior derivatization or sample pretreatment. An ion chromatography (IC) method was developed with an MS detector.| Market: | Environmental |
| Keywords: | drinking water, ESI, fluoroacetic acid, MSQ Plus, RFIC, Rodenticide, SIM, single quad MS |
| Matrix: | Water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.