Find methods for your needs
TN145: Determination of Total Inorganic Arsenic in Fruit Juice Using High-Pressure Capillary Ion Chromatography
Description
This application presents a method that detects and quantifies total inorganic arsenic (Arsenate As(V) and Arsenite As(III)) in fruit juices using a high-pressure capillary IC system with suppressed conductivity detection (CD), and with a high-resolution 4 µm particle anion–exchange column AS11-HC-4μm.| Market: | Food and Beverage |
| Keywords: | 4 μm Column, Apple juice, Arsenate, Arsenite, Conductivity Detection, Food and Beverage, High Resolution, HPIC, Organic acid, RFIC, Total Inorganic Arsenic |
| Matrix: | Juice |
For Research Use Only. Not for use in diagnostic procedures.