Find methods for your needs
AN144: Determination of Perchlorate in High Ionic Strength Fertilizer Extracts by Ion Chromatography.
Description
This application note provides a procedure for preparing aqueous leachates of fertilizers, inorganic salts, and other materials. The perchlorate anion in these extracts can be determined in 15 min by using a Thermo Scientific Dionex IonPac AS16 column, EG40-generated hydroxide eluent, a 500-μL injection, and suppressed conductivity detection. IonPac AS16 column is a high capacity, hydroxide-selective anion exchange column with ultra low hydrophobicity designed for the isocratic separation of polarizable anions including perchlorate, iodide, and thiosulfate.| Market: | Environmental |
| Keywords: | ED50 Conductivity Detector, EG40 Eluent Generator, Fertilizer extract, IonPac AS16, Perchlorate, RFIC-EG system, Suppressed Conductivity Detection |
| Matrix: | Chilean nitrate fertilizer extract |
| Author: | David H. Thomas and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.