Find methods for your needs
AN176: Determining Sub-ppb Perchlorate in Drinking Water Using Preconcentration/Matrix Elimination IC with Suppressed Conductivity Detection by U.S. EPA Method 314.1 Be the first to rate this application
determination perchlorate water
Description
In this application , we demonstrate the the determination of trace (< 1 ppb) perchlorate in drinking water samples and a simulated high-ionic-strength matrix.
Market: Environmental
Keywords: Confirmatory Method, Matrix Elimination, Preconcentration, Primary Method, Thermo Scientific Dionex IonPac AS16 Column, Thermo Scientific Dionex IonPac AS20 Column
Matrix: Drinking water
Author: Brian De Borba and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 1/29/2015.
For Research Use Only. Not for use in diagnostic procedures.