Find methods for your needs
AN220: Determination of Inorganic Anion Impurities in a Water-Insoluble Pharmaceutical by Ion Chromatography with Suppressed Conductivity Detection Be the first to rate this application
Description
Pharmaceutical impurities are categorized as organic, inorganic, or residual solvents. Inorganic impurities may be derived from the manufacturing process of bulk drugs. In most cases, these impurities should be removed or at least minimized in the final product. Therefore, the identification, quantification, and control of impurities are important during drug development in the pharmaceutical industry. In this Application Note, we demonstrate the development of an IC method for the determination of anionic impurities in a proprietary water-insoluble pharmaceutical.
Market: Pharma
Keywords: API, AS15, counter ion, Counterion, inorganic impurities, IonPac AS15, Pharmaceutical, AN 220, insoluble API, insoluble drug, AN220
Matrix: Water insoluble pharmaceutical
Author: Brian De Borba and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 4/23/2010.
For Research Use Only. Not for use in diagnostic procedures.