Find methods for your needs
AN175: Determination of Sulfate and Chloride in Ethanol by Ion Chromatography
Description
This application note describes the simplest, most direct method to measure mg/L concentrations of chloride and sulfate anions in ethanol. Sulfate and chloride are determined in 15 min by using IC with eluent generation and suppressed conductivity detection, on either an IonPac AS14A or IonPac AS18 column. The method is sensitive enough to monitor compliance with ASTM Specification D 4806. The methods described here were validated by Dionex during participation in the ASTM interlaboratory collaborative study to determine precision and bias for the new Standard Test Method.| Market: | Petrochem, Oil and Gas |
| Keywords: | ASTM Specification D 4806, Gasoline Blending, IonPac AS14A, IonPac AS18, Method WK9828, Method WK9829, RFIC, Spark Ignition Fuel |
| Matrix: | Ethanol |
For Research Use Only. Not for use in diagnostic procedures.