Find methods for your needs
Simple HILIC separation of arginine and glutamic acid without derivatization using HPLC-UV
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the analysis of two amino acids. The separation or arginine and glutamic acid was performed in less than 10 minutes on a Thermo Scientific Acclaim Mixed-Mode HILIC-1 HPLC column without the need for derivatization.| Market: | Metabolomics |
| Keywords: | Acclaim Mixed-Mode HILIC-1, Amino acids, Arginine, Glutamic acid, HPLC, UV detection |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.