Find methods for your needs
Glycans by UHPLC-CAD on Thermo Scientific Dionex UltiMate 3000 RSLC
Description
Glycans released from bovine fetuin by PNGase-F are separated by gradient elution on the Thermo Scientific Dionex GlycanPac AXR-1 column and detected by charged aerosol detection on the Corona Veo RS.Market: | BioPharma |
Keywords: | 2-AA labeling, 2-AB labeling, glycan profiling, protein characterization, Universal Detection |
Matrix: | Fetuin standard reconstituted in deionized water |
For Research Use Only. Not for use in diagnostic procedures.