Find methods for your needs
Determining Vicinal Diketones in Beer Using Valve-and-Loop Headspace Analysis
Description
The Thermo Scientific Triplus 300 Headspace Autosampler has been coupled to the Thermo Scientific Trace 1310 Gas Chromatograph for the analysis of 2,3-pentanedione and 2,3 butanedione in beer according to the method issued by the European Brewery Convention in 1999. One critical point of this method is the need to incubate the samples at 35°C. In this study the Triplus 300 HS has been successfully operated at a constant temperature of 35°C without any cryogenic device.Market: | Food and Beverage |
Keywords: | beer aroma, Flavor, food quality, Static Headspace, Triplus 300 HS, electron capture detector |
Matrix: | Beer |
For Research Use Only. Not for use in diagnostic procedures.