Find methods for your needs
Determination of Oxyhalides and Bromide in Drinking Water According to EPA Method 300.1
Description
Determination of oxyhalides and bromide in drinking water samples using EPA Method 300.1 on a Thermo Scientific Dionex IC-5000 Reagent-Free Ion Chromatography (RFIC) system.The anions were separated with a Thermo Scientific Dionex IonPac AS27 anion-exchange column using an electrolytically generated gradient KOH eluent.| Market: | Environmental |
| Keywords: | Bromate, Bromide, Chlorate, Chlorite, Dionex IonPac AS27, Disinfection Byproduct, Drinking Water, Environmental, EPA Method 300.1 (B), Ion Chromatography, Ion Chromatography (IC), Water analysis |
| Matrix: | Drinking water (spiked) |
For Research Use Only. Not for use in diagnostic procedures.