Find methods for your needs
AN1103: Determination of Total Nitrogen and Phosphorus in Wastewaters by Alkaline Persulfate Digestion Followed by IC
Description
This application demonstrates the simultaneous determination of total nitrogen (as nitrate-N) and total phosphorus (as phosphate-P) using a high-capacity hydroxide-selective Thermo Scientific™ Dionex™ IonPac™ AS19 Analytical Column with suppressed conductivity detection after alkaline persulfate digestion.| Market: | Environmental |
| Keywords: | Dionex IonPac AS19 Column, Eutrophication, Ion Chromatography (IC), Nutrient Pollution, Suppressed Conductivity Detection, Total Kjeldahl Nitrogen (TKN), Total Phosphorus, Total Nitrogen |
| Matrix: | Surface, Wastewaters |
| Author: | Brian De Borba, Richard F. Jack, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.