Find methods for your needs
Rapid assay for Rutoside Trihydrate in a solid dosage formulation using a Thermo Scientific Accucore aQ HPLC column
Description
The Thermo Scientific Ultimate 3000 RS system is applied for the assay analysis of Rutoside Trihydrate. The Thermo Scientific Accucore aQ column separated the rutoside trihydrate from the impurities present in a solid dosage formulation in under 15 minutes.| Market: | Pharma |
| Keywords: | Accucore aQ, Flavonoid, HPLC, Rutoside trihydrate, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.