Find methods for your needs
Rapid determination of Mometasone Furoate in formulation using a Thermo Scientific Acclaim Polar Advantage HPLC column
Description
The Thermo Scientific UltiMate 3000 system is applied for the rapid determination of mometasone furoate in formulation. The analysis is performed using the Thermo Scientific Acclaim Polar Advantage 2.2 µm column with a run time of less than three minutes.| Market: | Pharma |
| Keywords: | Acclaim Polar Advantage (PA), Glucocorticosteroid, HPLC, Mometasone furoate, Nasonex, UV detection |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.