Find methods for your needs
High resolution separation by GC of derivatized acylated sugars using a Thermo Scientific TRACE TR-1701 GC column. Be the first to rate this application
high resolution separation by gc derivatized acylated sugars using a thermo scientific trace tr1701 gc column
# Compound Name Compound Class RT (min)
1 Fructose Saccharides 16.00
2 Glucose anomers Saccharides 16.10
3 Glucose anomers Saccharides 17.00
Description
The Thermo Scientific TRACE GC Ultra is applied to the separation of derivatized acylated sugars using the Thermo Scientific TRACE TR-1701 column with FID detection
Market: Food and Beverage
Keywords: acylation, anomers, Derivatization reagent, fructose, glucose, N-methyl-bis(trifluoroacetamide), sugars, TR-1701
Matrix: Derivatization product
Uploaded on 10/29/2014.
For Research Use Only. Not for use in diagnostic procedures.