Find methods for your needs
A rapid method for determination of liposomal and non-liposomal (free) Doxorubicin from human plasma by SPE and LC-MS/MS using a Thermo Scientific Accucore C18 column.
Description
The Thermo Scientific Dionex UltiMate 3000 system was applied with the Thermo Scientific TSQ Vantage with a Thermo Scientific Accucore C18 2.6 μm column for the separation and determination of liposomal and non-liposomal (free) Doxorubicin from human plasma with MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore C18, Antineoplastic Antibiotic, daunorubicin, Doxorubicin, SOLA, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.