Find methods for your needs
An improved analysis of Haloacetic acids and Dalapon in water by GC-MS using a Thermo Scientific TraceGOLD TG-5SilMS with SafeGuard GC column
Description
The Thermo Scientific TRACE Ultra GC was used to demonstrate the performance and suitability of using an ultra low bleed sylarylene stationary phase (polarity similar to 5 % phenyl methylpolysiloxane) GC column with an integrated guard column for the identification of haloacetic acids and dalapon in water. This method can provide an alternative confirmation identification using EPA method 552.2.| Market: | Water Analysis |
| Keywords: | Dalapon, Derivatization, EPA method 552, Guard column, Haloacetic acids (HAA), Liquid-liquid extraction, TraceGOLD TG-5SilMS with SafeGuard |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.