Description
本方法与抑制电导离子色谱法相比较,选择性更好,抗干扰能力更强,检测结果更为可靠。本方法可为刑事侦查中草甘膦中毒等相关案件分析提供必要的论据。| Market: | Clinical Research |
| Keywords: | 离子色谱;电导;IonPac AS19;血液;草甘膦;质谱 |
| Matrix: | 血液 |
For Research Use Only. Not for use in diagnostic procedures.