Find methods for your needs
Separation of phenol and its chlorinated derivatives using reversed-phase HPLC-UV
Description
The testing here shows the advantages of using the Thermo Scientific Dionex UltiMate 3000 system with the Thermo Scientific Accucore aQ 2.6 µm column for the separation of phenol and its chlorinated derivatives with UV detection at 270 nm and a total run time of 25 minutes.| Market: | Environmental |
| Keywords: | Accucore aQ, Chlorinated phenol derivatives, HPLC, Phenol, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.