Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the determination of metformin. The separation is performed on a Thermo Scientific Syncronis HILIC 5 µm column, with UV detection at 225 nm and a total run time of 6 minutes.| Market: | Pharma |
| Keywords: | HILIC, HPLC, Metformin, Syncronis HILIC, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.