Find methods for your needs
AN1083: Determination of Myo-Inositol (Free and Bound as Phosphatidylinositol) in Infant Formula and Adult Nutritionals Be the first to rate this application
determination myoinositol
Description
This application evaluates and describes an HPAE-PAD method combined with the column-switching technique described in AOAC Official Method 2011.18, for the determination of free and bound myo-inositol in infant formula and adult nutritionals.
Market: Food and Beverage
Keywords: Au Disposable Working Electrode, Column Switching, Dionex CarboPac MA1 Column, Dionex CarboPac PA1 Column, Dionex OnGuard II RP Cartridge, TN 20, TN 70671, TN20, TN70671, AN 1083
Matrix: Water
Author: Lillian Chen, Brian De Borba, and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 10/14/2014.
For Research Use Only. Not for use in diagnostic procedures.