Find methods for your needs
Reaction monitoring separation of atorvastatin and its impurities using a PFP core enhanced technology HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the reaction monitoring separation of avorvastatin and its impurities. The separation was performed on a Thermo Scientific Accucore PFP column, with UV detection at 248 nm and a total run time of 18 minutes.| Market: | Pharma |
| Keywords: | Accucore PFP, Atorvastatin, HPLC, Statins, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.