Find methods for your needs
AN54: Determination of Total and Free Sulfite in Foods and Beverages
Description
In the food and beverage industries, sulfing agents have been used as preservatives to prevent microbial spoiling and browning reactions in a wide variety of food and beverage products. The U.S. Food and Drug Administration (FDA) requires labiling of any food or beverage containing a sulfite concentration > 10 ppm. This application describes an imporved ion-exclusion chromatography with pulsed amperometric detection method for the determination of total and free sulfite in foods and beverages.| Market: | Food and Beverage |
| Keywords: | Dionex IonPac ICS AS1 Column, Disposable Platinum Electrode, Electrochemical Detection, Ion-Exclusion Chromatography, Pulsed Amperometric Detection |
| Matrix: | Water |
| Author: | Lillian Chen, Brian De Borba, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.