Find methods for your needs
Neuroactive Amino Acids Be the first to rate this application
neuroactive amino acids
Description
The Thermo Scientific Dionex UltiMate system is applied for the analysis of amino acids. These are the most abundant neurochemicals in the brain. There are both excitatory (Glutamate) and inhibitory (GABA) amino acids that act as neurotransmitters. Analysis of amino acids requires the preparation of derivatives to achieve sufficient sensitivity for analysis. The OPA/β-ME derivative of an amino acid generates compounds which are electroactive.
Market: BioPharma
Keywords: Amino acids
Matrix: Brain extracellular fluid
Uploaded on 10/10/2014.
For Research Use Only. Not for use in diagnostic procedures.