Find methods for your needs
Rapid separation of high and low molecular weight peptides using a solid core C18 HPLC column
Description
The separation of peptides was successfully achieved on the Thermo Scientific Accucore C18 HPLC column. Basline resolution was achieved between high and low molecular weight peptides with a simple gradient method with a run time of 10 minutes| Market: | BioPharma |
| Keywords: | Accucore C18, Glycyl-L-Tyrosine, HPLC, Insulin, Leucine enkephalin, Methionine enkephalin, Peptides, Ribonuclease, UV detection, Val-Tyr-Val |
| Matrix: | N/A |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.