Find methods for your needs
Fast analysis of fexofenadine and pseudoephedrine using a Thermo Scientific Acclaim Polar Advantage (PA) HPLC column Be the first to rate this application
1
2
3
4
00.81.62.43.24Time [min]-20020406080100120140Response [mAU]
# Compound Name Compound Class RT (min)
1 Artifact Artifact 0.60
2 Pseudoephedrine Decongestant 0.98
3 Artifact Artifact 1.75
4 Fexofenadine Antihistamine 2.85
Description
The Thermo Scientific Acclaim Polar Advantage (PA) HPLC column is used to separate fexofenadine and pseudoephedrine formulation in less than four minutes.
Market: Pharma
Keywords: Acclaim Polar Advantage (PA), Fexofenadine, HPLC, Pseudoephedrine, UV detection
Matrix: Tablet
Uploaded on 2/17/2010.
For Research Use Only. Not for use in diagnostic procedures.