Find methods for your needs
Improved separation of resveratrol and related compounds in red wine using a Thermo Scientific Acclaim PolarAdvantage II (PA2) HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of resveratrol and related compounds in red wine. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II (PA2) HPLC column, with UV detection at 310 nm.Market: | Food and Beverage |
Keywords: | Acclaim PolarAdvantage II (PA2), HPLC, Polydatin, Pterostilbene, Red wine, Resveratrol, UltiMate 3000 RSLC, UV detection |
Matrix: | Red wine |
For Research Use Only. Not for use in diagnostic procedures.