Find methods for your needs
Fast analysis of blue dyes in breakfast cereal using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of FD&C blue #1 and #2 dyes in blueberry-flavored breakfast cereal. The separation is performed on a Thermo Scientific Acclaim 120 C18 column. The cereal sample was ground, extracted and filtered prior to analysis.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Breakfast cereal, Dyes, FD&C blue, Filter, Food, HPLC, UV |
| Matrix: | Cereal |
For Research Use Only. Not for use in diagnostic procedures.