Find methods for your needs
Fast analysis of vitamins D2 & D3 on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column in HILIC mode
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the separation of vitamin D2 and D3, with UV detection at 285nm. The analysis was performed in HILIC mode.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, HILIC, HPLC, UV detection, Vitamin D2, Vitamin D3, Vitamins |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.