Find methods for your needs
An improved analysis of alkaloids in goldenseal root on a Thermo Scientific Acclaim 120 C18 HPLC column with Thermo Scientific Dionex ASE Accelerated Solvent Extractor Be the first to rate this application
an improved analysis alkaloids goldenseal root on a thermo scientific acclaim 120 c18 hplc column with thermo scientific dionex ase accelerated solvent extractor
Description
The alkaloids present in goldenseal root were extracted using a Thermo Scientific Dionex ASE Accelerated Solvent Extractor system before separation by HPLC with UV detection. The separation of hydrastine and berberine was performed in less than 10 minutes on a Thermo Scientific Acclaim 120 C18 HPLC column.
Market: Food and Beverage
Keywords: Accelerated Solvent Extractor, Acclaim 120 C18, Berberine, Goldenseal root, HPLC, Hydrastine, UV detection
Matrix: Goldenseal root
Uploaded on 7/18/2014.
For Research Use Only. Not for use in diagnostic procedures.