Find methods for your needs
Accurate determination of acrylic acid oligomers on a Thermo Scientific Acclaim Organic Acid (OA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of acrylic acid oligomers. The separation is performed on a Thermo Scientific Organic Acid HPLC column, with UV detection at 210 nm.Market: | Material Science |
Keywords: | Acclaim Organic Acid, Acrylic acid oligomers, HPLC, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.