Find methods for your needs
Accelerated assays of vanilla extract on a Thermo Scientific Acclaim 120 C18 RSLC column
Description
The AOAC Official Method 990.25 takes over 1 hour to run. In this work the Thermo Scientific Acclaim RSLC C18 column has improved selectivity and a much shorter run time. Using well-known geometric scaling rules, the assay can be accelerated sixfold. Doubling the flow rate accelerates the analysis 12-fold, with no sacrifice in the quality of the separation.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, AOAC Official Method 990.25, HPLC, p-Hydroxybenzaldehyde, p-Hydroxybenzoic acid, UV detection, Vanilla extract, Vanillic acid, Vanillin |
| Matrix: | Vanilla extract |
For Research Use Only. Not for use in diagnostic procedures.