Find methods for your needs
Improved analysis of antioxidants from artichoke on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column
Description
The Thermo Scientific Dionex system is applied for the analysis of antioxidants from artichoke. The separation was performed in less than 25 minutes on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column, with UV detection.| Market: | Food and Beverage |
| Keywords: | 1,5-Dicaffeoylquinic acid, 3-Caffeoylquinic acid, Accliam PolarAdvantage II (PA2), Antioxidants, Artichoke, HPLC |
| Matrix: | Artichoke |
For Research Use Only. Not for use in diagnostic procedures.