Find methods for your needs
Fast separation of microcystins on a Thermo Scientific Acclaim RSLC C18 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of microcystins. The separation is performed on a Thermo Scientific Acclaim RSLC 120 C18 2.2 µm column, with UV detection at 238 nm. While the standard method takes about an hour to run, a straightforward conversion to rapid separation LC (RSLC) using the Acclaim RSLC 120 C18 2.2 µm column shortens the analysis time to six minutes.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, HPLC, Microcystins, UltiMate 3000 RSLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.