Find methods for your needs
Fast analysis of barbiturates using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific Dionex HPLC system is applied for the analysis of barbiturates. The separation is performed using a Thermo Scientific Acclaim 120 C18 column, with UV detection at 210nm.| Market: | Clinical Research |
| Keywords: | Acclaim 120 C18, Amobarbital, Barbital, Barbiturates, Butabarbital, Depressant drugs, HPLC, Phenobarbital, Secobarbital, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.