Find methods for your needs
Improved determination of diisocyanates by OSHA Method 42 on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column Be the first to rate this application
improved determination diisocyanates by osha method 42 on a thermo scientific acclaim polaradvantage ii pa2 column
Description
The Thermo Scientific Dionex UltiMate LPG3400 HPLC system is applied for the determination of diisocyanates by OSHA method 42. The Thermo Scientific Acclaim PA2 column is capable of separating the isocyanate derivates from this complicated matrix.
Market: Material Science
Keywords: 1,6-Hexamethylenediisocyanate, 2,4-Toluenediisocyanate, 2,6-Toluenediisocyanate, Acclaim PA2, Diisocyanate, HPLC, OSHA Method 42
Matrix: N/A
Uploaded on 7/16/2014.
For Research Use Only. Not for use in diagnostic procedures.