Find methods for your needs
Comparison of catechin profiles of black and green teas using a Thermo Scientific Acclaim PolarAdvantage II (PA2) column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of catechins. Catechins found in black and green tea are separated in less than 15 minutes using the Thermo Scientific Acclaim PA2 3 µm column with UV detection at 280 nm.| Market: | Food and Beverage |
| Keywords: | 3,4,5-trihydroxy cinnamic acid, Acclaim PA2, Caffeine, Catechin, Catechin gallate, Epicatechin, Epicatechin gallate, Epigallocatechin, Epigallocatechin gallate, Gallic acid, Gallocatechin, Gallocatechin gallate, Theobromine |
| Matrix: | Black tea, Green |
For Research Use Only. Not for use in diagnostic procedures.