Find methods for your needs
Improved determination of catechins in mango and tea using a Thermo Scientific Acclaim PolarAdvantage II (PA2) column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of catechins. The separation of ten catechins was performed in 30 minutes on a Thermo Scientific Acclaim PA2 3 µm column with UV detection at 280 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim PA2, Caffeine, Catechin, Catechins, Epicatechin, Epicatechin gallate, Epigallocatechin, Epigallocatechin gallate, Gallic acid, Gallocatechin, Gallocatechin gallate, Green tea, HPLC, Mango, Theobromine |
| Matrix: | Green tea, Mango pulp |
For Research Use Only. Not for use in diagnostic procedures.