Find methods for your needs
Fast determination of 4,4'-methylene bisphenyl isocyanate (MDI) by OSHA Method 47 on a Thermo Scientific Acclaim PolarAdvantage II (PA2) column Be the first to rate this application
fast determination 44methylene bisphenyl isocyanate mdi by osha method 47 on a thermo scientific acclaim polaradvantage ii pa2 column
Description
In this work the suitability of the Thermo Scientific Acclaim PA2 5 µm column for OHSA method 47 for the testing of workplace air for contamination. With a total run time of 8 minutes the method allows the separation of the isocyantate derivates and the matrix from each other.
Market: Environmental
Keywords: 4,4'-Methylene Bisphenyl Isocyanate (MDI), Acclaim PolarAdvantage II (PA2), HPLC, OSHA Method 47, urethane polymers
Matrix: N/A
Uploaded on 7/14/2014.
For Research Use Only. Not for use in diagnostic procedures.