Find methods for your needs
Determination of N-Methylpyrrolidine in Cefepime Using a Reagent-Free Ion Chromatography System
Description
This application describes a cation-exchange chromatography method for the determination of N-methylpyrrolidine (NMP) in Cefepime using the Thermo Scientific Dionex IonPac CS17 column and suppressed conductivity detection.| Market: | Pharma |
| Keywords: | cation exchange, Degradation Products, Drug Substance, IonPac CS17, USP |
| Matrix: | Cefepime drug substance |
For Research Use Only. Not for use in diagnostic procedures.