Find methods for your needs
Rapid recovery of Nitrofuran antibiotics from pet food using a Thermo Scientific Acclaim 120 C18 RSLC column Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the rapid determination of Nitrofuran antibiotics from pet food. The separation is performed on a Thermo Scientific Acclaim 120 C18 RSLC column in under 2 mins, with UV detection at 365 nm.
Market: Food and Beverage
Keywords: Acclaim 120 C18, Furaltadone, Furazolidone, HPLC, Nitrofuran Antibiotics, Nitrofurantoin, Nitrofurazone, Pet Food, UV detection
Matrix: Dry pet food
Uploaded on 2/15/2010.
For Research Use Only. Not for use in diagnostic procedures.