Find methods for your needs
Rapid analysis of nicotine tartrate using the Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific Acclaim Trinity P1 column was used to separate nicotine and its counterion. The nicotine and tartrate were separated using a phosphate buffer system to permit isocratic elution and UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, Alkaloid, HPLC, Nicotine, Tartrate, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.