Find methods for your needs
Demonstration of EPA Method 555 to separate chlorinated acids in ground water using a Thermo Scientific Acclaim RSLC PolarAdvantage HPLC column Be the first to rate this application
Description
EPA Method 555 is a HPLC method for the determination of certain chlorinated acids in ground water and finished drinking water using reversed-phase columns and diode-array detection. In this example, a fast, high-resolution separation of chlorinated acids in ground water is achieved using the Thermo Scientific Acclaim RSLC PolarAdvantage 2.2 µm column with an analysis cycle time of about 12 minutes.
Market: Environmental
Keywords: 2-(2,4,5-trichlorophenoxy)propionic acid, 2,4,5-TP, 2,4-Dichlorophenoxyacetic acid (2,4-D), Acifluorfen, Bentazon, Chloramben, Chlorinated acids, Dicamba, Dichloroprop, EPA Method 555, HPLC, Picloram, UV detection
Matrix: Ground water
Uploaded on 6/19/2014.
For Research Use Only. Not for use in diagnostic procedures.