Find methods for your needs
Simultaneous analysis of the basic drug Trimipramine and its counterion using the Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column Be the first to rate this application
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 retains trimipramine, a hydrophobic base, by reversed-phase interaction, and maleate, the hydrophilic counterion, by anion-exchange interaction. In this way, it can eliminate a separate test for the two components.
Market: Pharma
Keywords: Acclaim Mixed-Mode WAX-1, HPLC, Trimipramine, UV detection
Matrix: N/A
Uploaded on 6/17/2014.
For Research Use Only. Not for use in diagnostic procedures.